Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-2-ethylhexyl Phthalate, MP Biomedicals

Catalog No. ICN206013 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206013 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206013 Supplier MP Biomedicals, Inc Supplier No. 0520601380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117-81-7
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
Synonym dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil
PubChem CID 8343
ChEBI CHEBI:17747
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Quantity 100 g
Formula Weight 390.56
Chemical Name or Material Di-2-ethylhexyl Phthalate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.