Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-2-ethylhexylamine, 99%

Catalog No. AC271960010
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC271960010 1 L
AC271962500 250 mL
AC271960025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC271960010 Supplier Thermo Scientific Chemicals Supplier No. 271960010
Only null left
Add to Cart

CAS: 106-20-7 | C16H35N | 241.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-20-7
Molecular Formula C16H35N
Molecular Weight (g/mol) 241.46
MDL Number MFCD00009489
InChI Key SAIKULLUBZKPDA-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl amine, di 2-ethylhexyl amine, diisooctylamine, 1-hexanamine, 2-ethyl-n-2-ethylhexyl, bis-2-ethylhexylamine, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, ccris 4619
PubChem CID 7791
IUPAC Name 2-ethyl-N-(2-ethylhexyl)hexan-1-amine
SMILES CCCCC(CC)CNCC(CC)CCCC

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 281.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4415 to 1.4435
Linear Formula (CH3(CH2)3CH(C2H5)CH2)2NH
Quantity 1 L
Beilstein 04,388
Specific Gravity 0.8
Solubility Information Solubility in water: <0.2g/L (20 c) in water. Other solubilities: soluble in hydrocarbons
Viscosity 3.7 mPa.s (20°C)
Formula Weight 241.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Di-2-ethylhexylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-372-4
RTECSNumber IH6825000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.