Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-2-pyridyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. C14071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14071G 1 g
C14075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14071G Supplier TCI America Supplier No. C14071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1659-31-0
Molecular Formula C11H8N2O3
Molecular Weight (g/mol) 216.20
MDL Number MFCD00191407
InChI Key GCSAXWHQFYOIFE-UHFFFAOYSA-N
Synonym Carbonic Acid Di-2-pyridyl Ester
PubChem CID 2757370
IUPAC Name bis(pyridin-2-yl) carbonate
SMILES O=C(OC1=CC=CC=N1)OC1=CC=CC=N1

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-2-pyridyl Carbonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.