Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-2-thienyl sulfide, 97%

Catalog No. AAH3328406
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH3328406

Supplier: Thermo Scientific Chemicals H3328406

Only null left
Add to Cart

CAS: 3988-99-6 | C8H6S3 | 198.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3988-99-6
Molecular Formula C8H6S3
Molecular Weight (g/mol) 198.32
MDL Number MFCD00052047
InChI Key AKYIWBKPINOXJY-UHFFFAOYSA-N
Synonym 2-2-thienylthio thiophene, thiophene, 2,2'-thiobis, thiophene,2,2'-thiobis, 2-2-thienylsulfanyl thiophene, 2-thiophen-2-ylsulfanyl thiophene, 2-thienyl sulfide, di-2-thienyl sulfide, 2,2'-thiobisthiophene, di 2-thienyl sulphide
PubChem CID 138094
IUPAC Name 2-thiophen-2-ylsulfanylthiophene
SMILES S(C1=CC=CS1)C1=CC=CS1

Specifications

Density 1.311
Boiling Point 102°C to 104°C (0.9 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.649
Quantity 5 g
Formula Weight 198.32
Percent Purity 97%
Chemical Name or Material Di-2-thienyl sulfide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.