Learn More
Aat Bioquest Di-4-ANEPPS CAS 90134-00-2 (5 mg)
SDP
Catalog No. 502250007
Encompass_Preferred
Potential-sensitive ANEP dyes are zwitterionic molecule exhibiting the most consistent potentiometric response in different cell and tissue types. This catalog is for 5 mg.
This item is not returnable. View return policy
Catalog No. 50-225-0007 Supplier Aat Bioquest Supplier No. 21498
Potential-sensitive ANEP dyes are zwitterionic molecule exhibiting the most consistent potentiometric response in different cell and tissue types. This catalog is for 5 mg.