Learn More
Biotium DI-4-ANEPPS
Supplier:  Biotium 61010
Encompass_Preferred
Di-4-ANEPPS is a fast-responding membrane potential dye. Changes in the membrane potential of the cell correlate with changes in the fluorescence excitation intensity of the dye. Properties: Ex/ Em(MeOH) = 496/705 nm; Orange red solid soluble in DMF, ethanol or DMSO; Store at 4°C and protect from light, especially in solution; C28H36N2O3S; MW: 48; [90134-00-2]
Catalog No. 50-196-4731