Learn More
Strem, An Ascensus Company CAS# 52462-29-0. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. 1G.
SDP
Catalog No. 5090111853
Encompass
CAS# 52462-29-0. 1G. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. Molecular Formula: C20H28Cl4Ru2. Molecular Weight: 612.39. Color/Form: brown pwdr. Strem#: 44-0190 www.strem.com/catalog/v3/44-0190/
Catalog No. 50-901-11853 Supplier Strem, An Ascensus Company Supplier No. 4401901G
CAS# 52462-29-0. 1G. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. Molecular Formula: C20H28Cl4Ru2. Molecular Weight: 612.39. Color/Form: brown pwdr. Strem#: 44-0190 www.strem.com/catalog/v3/44-0190/