Learn More
Strem, An Ascensus Company CAS# 12148-71-9. Di-μ-methoxobis(1,5-cyclooctadiene)diiridium(I), min. 98%. MFCD08459360. 2G
SDP
Catalog No. 5090113126
Encompass
CAS# 12148-71-9. 2G. Di-μ-methoxobis(1,5-cyclooctadiene)diiridium(I), min. 98%. MFCD08459360. Molecular Formula: C18H30Ir2O2. Molecular Weight: 662.86. Color/Form yellow pwdr. Strem#: 77-1100 www.strem.com/catalog/v3/77-1100/
Catalog No. 50-901-13126 Supplier Strem, An Ascensus Company Supplier No. 7711002G
CAS# 12148-71-9. 2G. Di-μ-methoxobis(1,5-cyclooctadiene)diiridium(I), min. 98%. MFCD08459360. Molecular Formula: C18H30Ir2O2. Molecular Weight: 662.86. Color/Form yellow pwdr. Strem#: 77-1100 www.strem.com/catalog/v3/77-1100/