Learn More
Medchemexpress LLC DI-591 | 2245887-38-9 | 98.1% | 585.80 | C31H47N5O4S | 10MM 1ML
SDP
Catalog No. 5000236677
Encompass_Preferred
DI-591 is a potent, high-affinity, cell-permeable inhibitor of the DCN1-UBC12 interaction that selectively inhibits cullin 3 neddylation. It is supplied for research use in both solution and solid forms and includes documented solubility and formulation protocols for in vitro and in vivo studies.
  • Binds DCN1 and DCN2 with Ki values of 12 nM and 10.4 nM.
  • Selective inhibition of cullin 3 neddylation.
  • Cell-permeable small molecule suitable for cellular assays.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (~98.1%) with supporting certificate of analysis.
  • Soluble in DMSO (12.5 mg/mL); recommended in vivo formulations provided.
Catalog No. 50-002-36677 Supplier Medchemexpress LLC Supplier No. HY12460210MM 1ML
DI-591 is a potent, high-affinity, cell-permeable inhibitor of the DCN1-UBC12 interaction that selectively inhibits cullin 3 neddylation. It is supplied for research use in both solution and solid forms and includes documented solubility and formulation protocols for in vitro and in vivo studies.
  • Binds DCN1 and DCN2 with Ki values of 12 nM and 10.4 nM.
  • Selective inhibition of cullin 3 neddylation.
  • Cell-permeable small molecule suitable for cellular assays.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (~98.1%) with supporting certificate of analysis.
  • Soluble in DMSO (12.5 mg/mL); recommended in vivo formulations provided.