Learn More
Medchemexpress LLC DI-591 | 2245887-38-9 | 98.4% | 585.80 | C31H47N5O4S | 50MG
SDP
Catalog No. 5000236743
Encompass_Preferred
DI-591 is a potent, cell-permeable small-molecule inhibitor of the DCN1-UBC12 interaction that blocks cullin 3 neddylation. It is intended for biochemical and cellular research where inhibition of neddylation and modulation of ubiquitin-pathway signaling are required.
Catalog No. 50-002-36743 Supplier Medchemexpress LLC Supplier No. HY12460250MG
DI-591 is a potent, cell-permeable small-molecule inhibitor of the DCN1-UBC12 interaction that blocks cullin 3 neddylation. It is intended for biochemical and cellular research where inhibition of neddylation and modulation of ubiquitin-pathway signaling are required.