Learn More
Medchemexpress LLC Di-591 | 2245887-38-9 | 98.1% | 585.80 g/mol | C31H47N5O4S | 25MG
SDP
Catalog No. 5000235513
Encompass_Preferred
DI-591 is a cell-permeable small-molecule inhibitor of the DCN1-UBC12 interaction that binds DCN1 and DCN2 with high affinity (Ki ≈ 12 nM and 10.4 nM) and selectively inhibits neddylation of cullin 3. It is supplied at research-grade purity for biochemical and cellular studies of neddylation and ubiquitin-proteasome pathway regulation.
  • High-affinity binding to DCN1 and DCN2 (Ki ≈ 12 nM, 10.4 nM)
  • Selective inhibition of cullin 3 neddylation
  • Cell-permeable for cellular assays
  • High purity (~98.1%) suitable for research use
  • Molecular weight 585.80 g/mol; formula C31H47N5O4S
  • Available in multiple milligram pack sizes for laboratory studies
Catalog No. 50-002-35513 Supplier Medchemexpress LLC Supplier No. HY12460225MG
DI-591 is a cell-permeable small-molecule inhibitor of the DCN1-UBC12 interaction that binds DCN1 and DCN2 with high affinity (Ki ≈ 12 nM and 10.4 nM) and selectively inhibits neddylation of cullin 3. It is supplied at research-grade purity for biochemical and cellular studies of neddylation and ubiquitin-proteasome pathway regulation.
  • High-affinity binding to DCN1 and DCN2 (Ki ≈ 12 nM, 10.4 nM)
  • Selective inhibition of cullin 3 neddylation
  • Cell-permeable for cellular assays
  • High purity (~98.1%) suitable for research use
  • Molecular weight 585.80 g/mol; formula C31H47N5O4S
  • Available in multiple milligram pack sizes for laboratory studies