Learn More
Medchemexpress LLC DI-591 | 2245887-38-9 | 98.4% | 585.80 | 10 MG
SDP
Catalog No. 5000365635
Encompass_Preferred
DI-591 is a potent, high-affinity, and cell-permeable inhibitor of the DCN1-UBC12 interaction. It selectively inhibits neddylation of cullin 3 with minimal effect on other cullin family members, making it an excellent probe compound for investigating the role of cullin 3 CRL in biological processes and human diseases. It also leads to NRF2 protein accumulation and transcriptional activation.
  • Binds to DCN1 and DCN2 with high affinity (Ki values of 12 nM and 10.4 nM respectively)
  • Shows >1000-fold selectivity over recombinant human DCN3-5 proteins
  • Disrupts the association of cellular DCN1 and UBC12 proteins
  • Increases mRNA levels of NQO1 and HO1
  • Used for research purposes only
Catalog No. 50-003-65635 Supplier Medchemexpress LLC Supplier No. 10MGHY124602
DI-591 is a potent, high-affinity, and cell-permeable inhibitor of the DCN1-UBC12 interaction. It selectively inhibits neddylation of cullin 3 with minimal effect on other cullin family members, making it an excellent probe compound for investigating the role of cullin 3 CRL in biological processes and human diseases. It also leads to NRF2 protein accumulation and transcriptional activation.
  • Binds to DCN1 and DCN2 with high affinity (Ki values of 12 nM and 10.4 nM respectively)
  • Shows >1000-fold selectivity over recombinant human DCN3-5 proteins
  • Disrupts the association of cellular DCN1 and UBC12 proteins
  • Increases mRNA levels of NQO1 and HO1
  • Used for research purposes only