Learn More
Biotium Di-8-ANEPPQ
Catalog No. 501964735
Encompass_Preferred
Di-8-ANEPPQ is a fast-responding membrane potential dye. It is more hydrophobic than Di-2-ANEPEQ and is useful for potential-sensitive retrograde labeling of neurons. Properties: Orange red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C38H59Br2N3; MW: 717.70
Catalog No. 50-196-4735 Supplier Biotium Supplier No. 61014
Di-8-ANEPPQ is a fast-responding membrane potential dye. It is more hydrophobic than Di-2-ANEPEQ and is useful for potential-sensitive retrograde labeling of neurons. Properties: Orange red solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C38H59Br2N3; MW: 717.70