Learn More
Biotium DI-8-ANEPPS
Catalog No. 501964733
Encompass_Preferred
ANEPPS dyes are fast responding membrane potential dyes. Both Di-4-ANEPPS and Di-8-ANEPPS are available. Di-4-ANEPPS has the problem of being internalized in cells over long-term, so it is mainly used for short-term experiments. However, because Di-8-ANEPPS is more lipophilic, the dye is better retained in the outer leaflet of cell plasma membranes and thus more suitable for long-term membrane potential studies. Di-8-ANEPPS is also more photostable and less phototoxic than Di-4-ANEPPS. Properties: Ex/Em (MeOH) = 498/713 nm; Orange red solid soluble in DMF, ethanol or DMSO; Store at 4°C and Protect from light, especially when in solution; C36H52N2O3S; MW:593; [157134-53-7]
Catalog No. 50-196-4733 Supplier Biotium Supplier No. 61012
Add to Cart
Edge
Add to Cart
ANEPPS dyes are fast responding membrane potential dyes. Both Di-4-ANEPPS and Di-8-ANEPPS are available. Di-4-ANEPPS has the problem of being internalized in cells over long-term, so it is mainly used for short-term experiments. However, because Di-8-ANEPPS is more lipophilic, the dye is better retained in the outer leaflet of cell plasma membranes and thus more suitable for long-term membrane potential studies. Di-8-ANEPPS is also more photostable and less phototoxic than Di-4-ANEPPS. Properties: Ex/Em (MeOH) = 498/713 nm; Orange red solid soluble in DMF, ethanol or DMSO; Store at 4°C and Protect from light, especially when in solution; C36H52N2O3S; MW:593; [157134-53-7]