Learn More
Medchemexpress LLC DI-82 | 1638148-50-1 | MFCD28386371 | 98.0% | 510.65 g·mol⁻¹ | C20H26N6O4S3 | 5 MG
SDP
Catalog No. 5000222272
Encompass_Preferred
DI-82 is a small-molecule inhibitor of deoxycytidine kinase (dCK) with reported biochemical potency and observed antitumor activity. It is supplied for research use in biochemical assays and preclinical studies as a powder form for laboratory workflows.
  • Potent dCK inhibition: IC50app 27.8 nM; Kiapp 9.2 nM.
  • High purity (98.0%) suitable for research applications.
  • Soluble in DMSO at 120 mg/mL; ultrasonic assistance may be needed.
  • Provided as a powder with recommended storage: -20°C for powder; -80°C for long-term storage in solvent.
  • Intended for in vitro studies and preclinical antitumor research.
Catalog No. 50-002-22272 Supplier Medchemexpress LLC Supplier No. HY1364995MG
DI-82 is a small-molecule inhibitor of deoxycytidine kinase (dCK) with reported biochemical potency and observed antitumor activity. It is supplied for research use in biochemical assays and preclinical studies as a powder form for laboratory workflows.
  • Potent dCK inhibition: IC50app 27.8 nM; Kiapp 9.2 nM.
  • High purity (98.0%) suitable for research applications.
  • Soluble in DMSO at 120 mg/mL; ultrasonic assistance may be needed.
  • Provided as a powder with recommended storage: -20°C for powder; -80°C for long-term storage in solvent.
  • Intended for in vitro studies and preclinical antitumor research.