Learn More
Medchemexpress LLC Di(adamantan-1-yl)phosphine | 131211-27-3 | 97.0% | 302.43 | 5 G
SDP
Catalog No. 5000395974
Encompass_Preferred
Di(adamantan-1-yl)phosphine is a biological molecule intended for research use only and not sold to patients. It appears as a white to off-white solid.
  • Store at 4°C, protected from light, and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
Catalog No. 50-003-95974 Supplier Medchemexpress LLC Supplier No. HYW0085485G
Di(adamantan-1-yl)phosphine is a biological molecule intended for research use only and not sold to patients. It appears as a white to off-white solid.
  • Store at 4°C, protected from light, and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen