Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Di(adamantan-1-yl)phosphine | 131211-27-3 | 98.0% | 302.43 | 1 G
SDP

Catalog No. 5000395860
Encompass_Preferred

Di(adamantan-1-yl)phosphine, also known as Di-1-adamantylphosphine, is a biological molecule classified as a biochemical assay reagent. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Chemical formula: C20H31P
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Catalog No. 50-003-95860 Supplier Medchemexpress LLC Supplier No. HYW0085481G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Di(adamantan-1-yl)phosphine, also known as Di-1-adamantylphosphine, is a biological molecule classified as a biochemical assay reagent. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Chemical formula: C20H31P
  • Ships at room temperature in continental US
  • Store at 4°C, protected from light and under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.