Learn More
Strem, An Ascensus Company CAS# 52462-29-0. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. 25G.
SDP
Catalog No. 501959566
Encompass
CAS# 52462-29-0. 25G. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. Molecular Formula: C20H28Cl4Ru2. Molecular Weight: 612.39. Color/Form: brown pwdr. Strem#: 44-0190 www.strem.com/catalog/v3/44-0190/
Catalog No. 50-195-9566 Supplier Strem, An Ascensus Company Supplier No. 44019025G
CAS# 52462-29-0. 25G. Di-μ-chlorobis[(p-cymene)chlororuthenium(II)], min. 98%. MFCD00064793. Molecular Formula: C20H28Cl4Ru2. Molecular Weight: 612.39. Color/Form: brown pwdr. Strem#: 44-0190 www.strem.com/catalog/v3/44-0190/