Learn More
Cayman Chemical di-coupled CBDP, Purity: ≥95%, CAS: 1260669-31-5, 5 mg
SDP
Catalog No. 502944578
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; a dialkylated form of CBDP; a byproduct in the synthesis of CBDP; intended for research and forensic applications
Catalog No. 50-294-4578 Supplier Cayman Chemical Supplier No. 413235 MG
An analytical reference standard that is structurally similar to known phytocannabinoids; a dialkylated form of CBDP; a byproduct in the synthesis of CBDP; intended for research and forensic applications