Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(ethylene glycol) hexyl ether

Catalog No. 4493931L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-939-31L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-939-31L Supplier Sigma Aldrich Supplier No. 4493931L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di(ethylene glycol) hexyl ether

Specifications

CAS 112-59-4
Melting Point -40°C
Density 0.935 g/mL (at 25°C)
Boiling Point 260°C
Molecular Formula C10H22O3
Refractive Index n20/D 1.4381 (literature)
Linear Formula CH3(CH2)5 OCH2CH2OCH2CH2OH
Quantity 1 L
MDL Number MFCD00010703
Synonym 2-(2-Hexyloxyethoxy)ethanol; C6E2; Hexyldiglycol
Molecular Weight (g/mol) 190.28
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.