Learn More
Strem, An Ascensus Company CAS# 1191-15-7. 100g. Di-i-butylaluminum hydride (20% in hexanes). MFCD00008928
SDP
Catalog No. 5090114045
Encompass
CAS# 1191-15-7. 100g. Di-i-butylaluminum hydride (20% in hexanes). MFCD00008928. Molecular Weight: 142.22. Molecular Formula: (C4H9)2AlH. Color/form: colorless liq. Strem# 93-1355. http://www.strem.com/catalog/v/93-1355/
Catalog No. 50-901-14045 Supplier Strem, An Ascensus Company Supplier No. 931355100G
CAS# 1191-15-7. 100g. Di-i-butylaluminum hydride (20% in hexanes). MFCD00008928. Molecular Weight: 142.22. Molecular Formula: (C4H9)2AlH. Color/form: colorless liq. Strem# 93-1355. http://www.strem.com/catalog/v/93-1355/