Learn More
Cell Signaling Technology Di-Methyl-Histone H3 (Lys4) Matched Antibody Pair, Human, ELISA, 1 Kit
SDP
Catalog No. 503133149
Encompass
The Di-Methyl-Histone H3 (Lys4) Matched Antibody Pair is ideal for use with immunoassay technologies and high-throughput ELISA platforms requiring specialized or custom antibody labeling. This kit contains capture and detection antibodies that bind to non-overlapping epitopes, supporting conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with high-throughput platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as a kit containing 100 μg each of Histone H3 (D2B12) and Di-Methyl-Histone H3 (Lys4) (C64G9) rabbit monoclonal antibodies.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)) without BSA or azide.
  • Validated for human species reactivity.
  • Store at -20°C; aliquot into single-use vials to avoid multiple freeze/thaw cycles.
Catalog No. 50-313-3149 Supplier Cell Signaling Technology Supplier No. 96365S
The Di-Methyl-Histone H3 (Lys4) Matched Antibody Pair is ideal for use with immunoassay technologies and high-throughput ELISA platforms requiring specialized or custom antibody labeling. This kit contains capture and detection antibodies that bind to non-overlapping epitopes, supporting conjugation methods including fluorophores, lanthanides, biotin, and beads.
  • Compatible with high-throughput platforms including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Supplied as a kit containing 100 μg each of Histone H3 (D2B12) and Di-Methyl-Histone H3 (Lys4) (C64G9) rabbit monoclonal antibodies.
  • Formulated in 1X PBS (10 mM Na2HPO4, 3 mM KCl, 2 mM KH2PO4, and 140 mM NaCl (pH 7.8)) without BSA or azide.
  • Validated for human species reactivity.
  • Store at -20°C; aliquot into single-use vials to avoid multiple freeze/thaw cycles.