Learn More
BIOHIPPO Di-Methyllysine rAb W-R22
SDP
Catalog No. 5000975576
Encompass
U-Blot Pan-Di-Methyllysine Rabbit mAb-BSA and Azide free is a Rabbit-derived recombinant monoclonal antibody (clone W-R22) that specifically recognizes Di-Methyllysine a lysine di-methylation modification at -amino group (N ) of the lysine (Lys K) side chain It shows reactivity with Human Mouse Rat Validated applications include WB Recommended dilutions WB 11000 Purified by protein a Di-Methyllysine (Kme2) is a crucial reversible post-translational modification of proteins which occurs on the -amino group of lysine residues including symmetric and asymmetric dimethylation forms It is widely distributed in histones and non-histone proteins and participates in regulating chromatin remodeling gene transcription RNA splicing DNA damage repair cell cycle progression and signal transduction Supplied as 100 uL
Catalog No. 50-009-75576 Supplier BIOHIPPO Supplier No. BHA21821333
U-Blot Pan-Di-Methyllysine Rabbit mAb-BSA and Azide free is a Rabbit-derived recombinant monoclonal antibody (clone W-R22) that specifically recognizes Di-Methyllysine a lysine di-methylation modification at -amino group (N ) of the lysine (Lys K) side chain It shows reactivity with Human Mouse Rat Validated applications include WB Recommended dilutions WB 11000 Purified by protein a Di-Methyllysine (Kme2) is a crucial reversible post-translational modification of proteins which occurs on the -amino group of lysine residues including symmetric and asymmetric dimethylation forms It is widely distributed in histones and non-histone proteins and participates in regulating chromatin remodeling gene transcription RNA splicing DNA damage repair cell cycle progression and signal transduction Supplied as 100 uL