Learn More
BIOHIPPO Di-Methyllysine Rabbit mAb
SDP
Catalog No. 5000975466
Encompass
U-Blot Pan-Di-Methyllysine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Di-Methyllysine a lysine di-methylation modification at -amino group (N ) of the lysine (Lys K) side chain Di-Methyllysine (Kme2) is a crucial reversible post-translational modification of proteins which occurs on the -amino group of lysine residues including symmetric and asymmetric dimethylation forms It is widely distributed in histones and non-histone proteins and participates in regulating chromatin remodeling gene transcription RNA splicing DNA damage repair cell cycle progression and signal transduction Aberrant lysine dimethylation is closely associated with tumorigenesis inflammation neurodegeneration and metabolic disorders serving as a vital biomarker for epigenetic and pathological research Supplied as 100 uL
Catalog No. 50-009-75466 Supplier BIOHIPPO Supplier No. BHA21821411
U-Blot Pan-Di-Methyllysine Rabbit mAb is a Rabbit-derived monoclonal antibody that specifically recognizes Di-Methyllysine a lysine di-methylation modification at -amino group (N ) of the lysine (Lys K) side chain Di-Methyllysine (Kme2) is a crucial reversible post-translational modification of proteins which occurs on the -amino group of lysine residues including symmetric and asymmetric dimethylation forms It is widely distributed in histones and non-histone proteins and participates in regulating chromatin remodeling gene transcription RNA splicing DNA damage repair cell cycle progression and signal transduction Aberrant lysine dimethylation is closely associated with tumorigenesis inflammation neurodegeneration and metabolic disorders serving as a vital biomarker for epigenetic and pathological research Supplied as 100 uL