Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-mu-chlorobis(2'-amino-1,1'-biphenyl-2-yl-C,N)dipalladium(II) 97.0+%, TCI America™
SDP

Catalog No. D41911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D41911G Supplier TCI America Supplier No. D41911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 847616-85-7
Molecular Formula C24H26Cl2N2Pd2+2
Molecular Weight (g/mol) 626.226
InChI Key FOAKXRLDQVUUEH-UHFFFAOYSA-N
PubChem CID 131674811
IUPAC Name dichloranium;palladium;2-phenylaniline
SMILES C1=CC=C(C=C1)C2=CC=CC=C2N.C1=CC=C(C=C1)C2=CC=CC=C2N.[ClH2+].[ClH2+].[Pd].[Pd]

Specifications

Melting Point 207°C
Quantity 1 g
Formula Weight 620.18
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis(2′-amino-1,1′-biphenyl-2-yl-C,N)dipalladium(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.