Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-mu-chlorobis[2-[(dimethylamino)methyl]phenyl-C,N]dipalladium(II) 97.0+%, TCI America™
SDP

Catalog No. D38471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38471G Supplier TCI America Supplier No. D38471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18987-59-2
Molecular Formula C18H26Cl2N2Pd2+2
Molecular Weight (g/mol) 554.16
MDL Number MFCD00167386
InChI Key DOROIKKMUTXYAD-UHFFFAOYSA-L
PubChem CID 131674728
IUPAC Name N,N-dimethyl-1-phenylmethanamine;palladium(2+);dichloride
SMILES CN(C)CC1=CC=CC=C1.CN(C)CC1=CC=CC=C1.[Cl-].[Cl-].[Pd+2].[Pd+2]

Specifications

Melting Point 176°C
Quantity 1 g
Formula Weight 552.14
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis[2-[(dimethylamino)methyl]phenyl-C,N]dipalladium(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.