Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-Mu-Chlorotetraethylene Dirhodium(I)

Catalog No. 6567631G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-676-31G 1 g
65-676-3100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 65-676-31G Supplier Sigma Aldrich Supplier No. 6567631G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Precursor to Rh(I) norbornadiene catalyst used in an asymmetric 1,4-addition of arylboronic acids to cyclic enones.

Specifications

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Linear Formula C8H16Cl2Rh2
MDL Number MFCD00013206
Quantity 1 g
Synonym Bis[chlorobis(ethylene)rhodium]; Bis[chlorodiethylenerhodium]; Chlorobis(ethylene)rhodium dimer; Dichlorotetra(ethene)dirhodium; Dichlorotetrakis(ethene)dirhodium; Diethylenerhodium chloride dimer; Divinylrhodium chloride dimer; Tetrakis(ethylene)dichlorodirhodium; [Rh(C2H4)2Cl]2
Molecular Weight (g/mol) 388.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.