Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-alkyldimethylammonium Chloride (mixture) 95.0+%, TCI America™
SDP

Catalog No. D163025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D163025G 25 g
D1630500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D163025G Supplier TCI America Supplier No. D163025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61789-80-8

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3077
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-n-alkyldimethylammonium Chloride (mixture)
EINECSNumber (2)-0184&(9)-1971
RTECSNumber UZ2997000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.