Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-amyl Ketone, MP Biomedicals

Catalog No. ICN209760 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN209760 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN209760 Supplier MP Biomedicals, Inc Supplier No. 0520976025

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 927-49-1
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
InChI Key ZPQAKYPOZRXKFA-UHFFFAOYSA-N
Synonym 6-undecanone, dipentyl ketone, amyl ketone, diamyl ketone, pentyl ketone, 6-oxoundecane, 6undecanone, di-n-amyl ketone, ketones, c11, c11 ketones
PubChem CID 13561
IUPAC Name undecan-6-one
SMILES CCCCCC(=O)CCCCC

Specifications

Quantity 100 g
Formula Weight 170.3
Chemical Name or Material Di-n-amyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.