Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyl carbonate

Catalog No. AC463880050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463880050 5 g
AC463880250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463880050 Supplier Thermo Scientific Chemicals Supplier No. 463880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 542-52-9 | C9H18O3 | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-52-9
Molecular Formula C9H18O3
Molecular Weight (g/mol) 174.24
InChI Key QLVWOKQMDLQXNN-UHFFFAOYSA-N
IUPAC Name dibutyl carbonate
SMILES CCCCOC(=O)OCCCC

Specifications

Density 0.9200 g/mL
Quantity 5 g
Specific Gravity 0.92
Formula Weight 174.24
Chemical Name or Material Di-n-butyl carbonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Causes serious eye damage.

Harmful to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 208-816-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.