Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyl maleate, 96%, Thermo Scientific™

Catalog No. AAH59123K1
Change view
Click to view available options
Quantity:
0.25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59123K1 0.25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH59123K1 Supplier Thermo Scientific Chemicals Supplier No. H59123K1

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-76-0
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00009447,MFCD00065141
InChI Key JBSLOWBPDRZSMB-FPLPWBNLSA-N
Synonym dibutyl maleate, butyl maleate, staflex dbm, rc comonomer dbm, maleic acid, dibutyl ester, di-n-butyl maleate, 2-butenedioic acid z-, dibutyl ester, 2-butenedioic acid, dibutyl ester, maleic acid dibutyl ester, 2-butenedioic acid 2z-, dibutyl ester
PubChem CID 5271569
IUPAC Name dibutyl (Z)-but-2-enedioate
SMILES CCCCOC(=O)\C=C/C(=O)OCCCC

Specifications

Density 0.988
Boiling Point 281°C
Flash Point 138°C (280°F)
Refractive Index 1.445
Linear Formula CH3(CH2)3OCOCH=CHCOO(CH2)3CH3
Quantity 0.25 L
Beilstein 1726634
Formula Weight 228.28
Percent Purity 96%
Chemical Name or Material Di-n-butyl maleate
Hazard Category H317-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-328-4
RTECSNumber ON0875000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.