Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butyl phthalate, 99%, Thermo Scientific Chemicals

Catalog No. aaa1325736
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1325736 500 g
AAA132570E 2500 g
AAA132570C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1325736 Supplier Thermo Scientific Chemicals Supplier No. A1325736
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a general purpose monomeric phthalate with excellent solubilizing characteristics. It is commonly used as an additive to adhesives and printing inks. It also finds utility in shower curtains, raincoats and vinyl coated fabrics.

Solubility
Soluble in water. (0.4 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-74-2
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD00009441
InChI Key DOIRQSBPFJWKBE-UHFFFAOYSA-N
Synonym dibutyl phthalate, di-n-butyl phthalate, n-butyl phthalate, butyl phthalate, celluflex dpb, elaol, genoplast b, palatinol c, polycizer dbp, unimoll db
PubChem CID 3026
ChEBI CHEBI:34687
IUPAC Name 1,2-dibutyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC

Specifications

Melting Point -35°C
Density 1.045
Boiling Point 339°C to 340°C
Flash Point 171°C (340°F)
Odor Odorless
Refractive Index 1.492
Quantity 500 g
UN Number UN3082
Beilstein 1914064
Merck Index 14,3035
Solubility Information Soluble in water. (0.4g/L)
Formula Weight 278.35
Percent Purity 99%
Chemical Name or Material Di-n-butyl phthalate
Show More Show Less
Hazard Category H336-H360Df-H373
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-557-4
RTECSNumber TI0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.