Learn More
Di-n-butylamine, 98+%
Description
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.
Solubility
Miscible with water, ether, ethanol and acetone.
Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.

Specifications
Specifications
Melting Point | -62°C |
Density | 0.767 |
Boiling Point | 159°C to 160°C |
Flash Point | 39°C (102°F) |
Odor | Amine-like |
Assay Percent Range | ≥98% |
Refractive Index | 1.4169 |
Linear Formula | [CH3(CH2)3]2NH |
Quantity | 2500 mL |
UN Number | UN2248 |
Show More |
Safety and Handling
Hazard Category | H226-H301+H311-H314-H330 |
Hazard Statement | GHS H Statement H226-H302-H312-H332 Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DI-N-BUTYLAMINE |
EINECSNumber | 203-921-8 |
RTECSNumber | HR7780000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.