Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-butylgermanium dichloride, 97%

Catalog No. AAA1713814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1713814 25 g
AAA1713806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1713814 Supplier Thermo Scientific Chemicals Supplier No. A1713814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4593-81-1 | C8H18Cl2Ge | 257.762 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4593-81-1
Molecular Formula C8H18Cl2Ge
Molecular Weight (g/mol) 257.762
MDL Number MFCD00013590
InChI Key VKMPTHGEGBKPBQ-UHFFFAOYSA-N
Synonym di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4
PubChem CID 20723
IUPAC Name dibutyl(dichloro)germane
SMILES CCCC[Ge](CCCC)(Cl)Cl

Specifications

Density 1.208
Boiling Point 241°C to 243°C
Flash Point 107°C (225°F)
Refractive Index 1.473
Linear Formula [CH3(CH2)3]2GeCl2
Quantity 25 g
UN Number UN2922
Sensitivity Moisture sensitive
Formula Weight 257.73
Percent Purity 97%
Chemical Name or Material Di-n-butylgermanium dichloride
Show More Show Less
Hazard Category H301+H311+H331-H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 224-986-9
RTECSNumber LY5020000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.