Learn More
Strem, An Ascensus Company CAS# 1067-33-0. 100g. Di-n-butyltin diacetate, min. 96%. MFCD00008697
SDP
Supplier:  Strem, An Ascensus Company 935007100G
Encompass
CAS# 1067-33-0. 100g. Di-n-butyltin diacetate, min. 96%. MFCD00008697. Molecular Weight: 351.01. Molecular Formula: (C4H9)2Sn(OOCCH3)2. Color/form: colorless liq. Strem# 93-5007. http://www.strem.com/catalog/v/93-5007/
Catalog No. 50-901-15546