Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-decyl dimethylammonium bromide, 80% aq. gel

Catalog No. AAB2483236
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAB2483236

Supplier: Thermo Scientific Chemicals B2483236

Only null left
Add to Cart

CAS: 2390-68-3 | C22H48BrN | 406.537 g/mol

Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-68-3
Molecular Formula C22H48BrN
Molecular Weight (g/mol) 406.537
MDL Number MFCD00012194
InChI Key UMGXUWVIJIQANV-UHFFFAOYSA-M
Synonym didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide
PubChem CID 16957
IUPAC Name didecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-]

Specifications

Refractive Index 1.457
Linear Formula [CH3(CH2)9]2N(CH3)2Br
Quantity 500 g
Solubility Information Soluble in methanol.
Formula Weight 406.54
Chemical Name or Material Di-n-decyl dimethylammonium bromide, 80% aq. gel
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 219-234-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.