Learn More
Medchemexpress LLC Di-n-dodecyl phthalate-3,4,5,6-d4 | 2708280-31-1 | MFCD28009755 | 98.0% | 506.79 | C32H50D4O4 | 5 MG
SDP
Catalog No. 5000285279
Encompass_Preferred
Di-n-dodecyl Phthalate-3,4,5,6-d4 is a deuterium-labeled analogue of di-n-dodecyl phthalate designed as an isotope-labeled tracer or internal standard for analytical quantitation and research applications. It incorporates four deuterium atoms at the 3,4,5,6 positions and is supplied as a high-purity material with recommended cold storage for long-term stability.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High isotopic enrichment suitable for quantitative analysis.
  • Recommended storage: -20°C in pure form; -80°C for solutions for extended periods.
  • Supplied in small-mass quantities appropriate for analytical workflows.
  • For research use only; not validated for medical applications.
Catalog No. 50-002-85279 Supplier Medchemexpress LLC Supplier No. HYW353555S5MG
Di-n-dodecyl Phthalate-3,4,5,6-d4 is a deuterium-labeled analogue of di-n-dodecyl phthalate designed as an isotope-labeled tracer or internal standard for analytical quantitation and research applications. It incorporates four deuterium atoms at the 3,4,5,6 positions and is supplied as a high-purity material with recommended cold storage for long-term stability.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • High isotopic enrichment suitable for quantitative analysis.
  • Recommended storage: -20°C in pure form; -80°C for solutions for extended periods.
  • Supplied in small-mass quantities appropriate for analytical workflows.
  • For research use only; not validated for medical applications.