Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-octyl phthalate, MilliporeSigma™ Supelco™

Catalog No. 111020495 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0495 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0495 Supplier MilliporeSigma Supelco Supplier No. 31301250MG
Add to Cart
Add to Cart

Di-n-octyl phthalate (DNOP) is a mixed component of C6-C10 linear-chain phthalates which has the ability to impart flexibility in plastics and rubber materials. It is also used as plasticizer in PVC used in flooring and carpet tiles.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 117-84-0
Density 0.980 g/mL (at 20°C)
Flash Point 109°C
Physical Form Neat
Linear Formula C24H38O4
MDL Number MFCD00015292
Synonym DNOP; Phthalic acid di-n-octyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 390.56
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.