Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-n-octyl Sebacate 95.0+%, TCI America™
SDP

Catalog No. S002825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S002825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S002825ML Supplier TCI America Supplier No. S002825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14491-66-8
Molecular Formula C26H50O4
Synonym Decanedioic Acid Di-n-octyl Ester, Sebacic Acid Di-n-octyl Ester, Di-n-octyl Decanedioate

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 mL
Formula Weight 426.68
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Di-n-octyl Sebacate
EINECSNumber (2)-0879&(2)-2569
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.