Learn More
Medchemexpress LLC Di(N-succinimidyl)adipate | 59156-70-6 | 95.0% | C14H16N2O8 | 250 MG
SDP
Catalog No. 5000352869
Encompass_Preferred
Di(N-succinimidyl)adipate is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Utilizes the ubiquitin-proteasome system for protein degradation
Catalog No. 50-003-52869 Supplier Medchemexpress LLC Supplier No. 250MGHY141099
Di(N-succinimidyl)adipate is an alkyl chain-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl chain-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Utilizes the ubiquitin-proteasome system for protein degradation