Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Di-o-cresyl phosphate | 35787-74-7 | 278.24 g/mol | C14H15O4P | 25 MG
SDP

Catalog No. 5000283874
Encompass_Preferred

Di-o-cresyl phosphate is an organophosphate compound used as a flame retardant and supplied as a research reagent. The compound has molecular formula C14H15O4P, molecular weight 278.24 g/mol, and CAS number 35787-74-7. Available pack sizes include 5 mg, 10 mg, 25 mg, 50 mg, and larger quantities on inquiry.

  • Organophosphate flame retardant.
  • CAS number: 35787-74-7.
  • Molecular formula: C14H15O4P.
  • Molecular weight: 278.24 g/mol.
  • Available pack sizes include 25 mg for small-scale studies.
  • Purity on product page; refer to manufacturer data sheet or SDS for purity and handling information.

Catalog No. 50-002-83874 Supplier Medchemexpress LLC Supplier No. HYW58592825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Di-o-cresyl phosphate is an organophosphate compound used as a flame retardant and supplied as a research reagent. The compound has molecular formula C14H15O4P, molecular weight 278.24 g/mol, and CAS number 35787-74-7. Available pack sizes include 5 mg, 10 mg, 25 mg, 50 mg, and larger quantities on inquiry.

  • Organophosphate flame retardant.
  • CAS number: 35787-74-7.
  • Molecular formula: C14H15O4P.
  • Molecular weight: 278.24 g/mol.
  • Available pack sizes include 25 mg for small-scale studies.
  • Purity on product page; refer to manufacturer data sheet or SDS for purity and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.