Learn More
Strem, An Ascensus Company CAS# 36042-94-1. 5G. Di-o-tolylchlorophosphine, min. 98%. MFCD04038733
SDP
Catalog No. 501963591
Encompass
CAS# 36042-94-1. 5G. Di-o-tolylchlorophosphine, min. 98%. MFCD04038733. Molecular Formula: (CH3C6H4)2PCl. Molecular Weight: 248.69. Color/Form: white to yellow hazy viscous liq. to solid. Stability: air sensitive, moisture sensitive. Strem# 15-1815. www.strem.com/catalog/v3/15-1815/
Catalog No. 50-196-3591 Supplier Strem, An Ascensus Company Supplier No. 1518155G
CAS# 36042-94-1. 5G. Di-o-tolylchlorophosphine, min. 98%. MFCD04038733. Molecular Formula: (CH3C6H4)2PCl. Molecular Weight: 248.69. Color/Form: white to yellow hazy viscous liq. to solid. Stability: air sensitive, moisture sensitive. Strem# 15-1815. www.strem.com/catalog/v3/15-1815/