Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-p-toluoyl-D-tartaric acid, 98%

Catalog No. AAL1322314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1322314 25 g
AAL1322322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1322314 Supplier Thermo Scientific Chemicals Supplier No. L1322314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32634-68-7
Molecular Formula C20H18O8
Molecular Weight (g/mol) 386.36
MDL Number MFCD00008552
InChI Key CMIBUZBMZCBCAT-HOTGVXAUSA-N
Synonym di-p-toluoyl-d-tartaric acid, +-di-1,4-toluoyl-d-tartaric acid, 2s,3s-2,3-bis 4-methylbenzoyl oxy succinic acid, unii-4480ma5qip, ditoluoyltartaric acid, +, +-dtta, butanedioic acid, 2,3-bis 4-methylbenzoyl oxy-, 2s,3s, 2s,3s-2,3-bis 4-methylbenzoyl oxy butanedioic acid, --di-p-toluoyl-l-trataric acid
PubChem CID 263211
SMILES CC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 169°C to 171°C
Beilstein 3225584
IUPAC Name (2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Formula Weight 386.36
Percent Purity 98%
Quantity 25 g
Optical Rotation +139° (c=1 in Ethanol)
Chemical Name or Material Di-p-toluoyl-D-tartaric acid
EINECSNumber 251-132-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.