Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-p-toluoyl-D-tartaric acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog No. AC148460050
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC148460050 5 g
AC148461000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148460050 Supplier Thermo Scientific Chemicals Supplier No. 148460050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71607-31-3
Molecular Formula C20H20O9
Molecular Weight (g/mol) 404.37
MDL Number MFCD00149567,MFCD00064440,MFCD00151034
InChI Key FOTRUJUPLHRVNU-UHFFFAOYNA-N
Synonym +-o,o'-di-p-toluoyl-d-tartaric acid monohydrate, 2r,3s-2,3-bis 4-methylbenzoyloxy butanedioic acid hydrate, 2s,3s-+-2,3-bis 4-methylbenzoyl oxy succinic acid monohydrate
PubChem CID 71299557
SMILES O.CC1=CC=C(C=C1)C(=O)OC(C(OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

Specifications

Melting Point 163.0°C to 165.0°C
Infrared Spectrum Authentic
Linear Formula [CH3C6H4CO2CH(CO2H)-]2·H2O
Quantity 5 g
Specific Rotation + 140.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and acetone
IUPAC Name 2,3-bis(4-methylbenzoyloxy)butanedioic acid hydrate
Formula Weight 404.38
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Di-p-toluoyl-D-tartaric acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.