Learn More
Medchemexpress LLC Di-p-tolyl phosphate | 843-24-3 | 278.24 | C14H15O4P | 25 MG
SDP
Catalog No. 5000285127
Encompass_Preferred
Di-p-Tolyl phosphate (CAS 843-24-3) is an organophosphate compound used as a flame retardant and as a reagent in biochemical and analytical applications. It is supplied in small laboratory quantities for research and testing; refer to the certificate of analysis for purity and storage recommendations.
  • Acts as an organophosphate flame retardant for polymers and formulations.
  • Used as a reagent or reference in biochemical and analytical assays.
  • Molecular formula C14H15O4P and molecular weight 278.24 g·mol⁻¹.
  • Supplied in small laboratory quantities suitable for research use.
  • Refer to the certificate of analysis for purity and recommended storage conditions.
Catalog No. 50-002-85127 Supplier Medchemexpress LLC Supplier No. HYW58755325MG
Di-p-Tolyl phosphate (CAS 843-24-3) is an organophosphate compound used as a flame retardant and as a reagent in biochemical and analytical applications. It is supplied in small laboratory quantities for research and testing; refer to the certificate of analysis for purity and storage recommendations.
  • Acts as an organophosphate flame retardant for polymers and formulations.
  • Used as a reagent or reference in biochemical and analytical assays.
  • Molecular formula C14H15O4P and molecular weight 278.24 g·mol⁻¹.
  • Supplied in small laboratory quantities suitable for research use.
  • Refer to the certificate of analysis for purity and recommended storage conditions.