Learn More
Medchemexpress LLC di-Pal-MTO | 1349197-90-5 | 98.4% | 917.27 | 5 MG
SDP
Catalog No. 5000362084
Encompass_Preferred
di-Pal-MTO is a palm oil-based lipid created by combining the anticancer agent mitoxantrone (MTO) with palmitoleic acid. When formulated into nanoparticles with mono-Pal-MTO in a 1:1 molar ratio, it demonstrates effective siRNA cell delivery and enhanced anticancer activity. This combination has been shown to significantly reduce tumor cell viability by 81% and tumor size by 83% in vitro, outperforming Lipofectamine 2000-mediated transfection.
  • Palm oil-based lipid, combining mitoxantrone (MTO) and palmitoleic acid.
  • Effective siRNA cell delivery, enhanced anticancer activity.
  • Forms nanoparticles (1:1 molar ratio with mono-Pal-MTO) for effective siRNA delivery.
  • Enhances in vitro antitumor activity, reducing tumor cell viability by 81% and tumor size by 83%.
Catalog No. 50-003-62084 Supplier Medchemexpress LLC Supplier No. HY1487025MG
di-Pal-MTO is a palm oil-based lipid created by combining the anticancer agent mitoxantrone (MTO) with palmitoleic acid. When formulated into nanoparticles with mono-Pal-MTO in a 1:1 molar ratio, it demonstrates effective siRNA cell delivery and enhanced anticancer activity. This combination has been shown to significantly reduce tumor cell viability by 81% and tumor size by 83% in vitro, outperforming Lipofectamine 2000-mediated transfection.
  • Palm oil-based lipid, combining mitoxantrone (MTO) and palmitoleic acid.
  • Effective siRNA cell delivery, enhanced anticancer activity.
  • Forms nanoparticles (1:1 molar ratio with mono-Pal-MTO) for effective siRNA delivery.
  • Enhances in vitro antitumor activity, reducing tumor cell viability by 81% and tumor size by 83%.