Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di-phenylethylammonium Lead(ii) Tetrabromide N = 1

Catalog No. 9109451G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-094-51G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-094-51G Supplier Sigma Aldrich Supplier No. 9109451G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This product is a violet emmiter for light emmiting diodes. It is solution processable in DMF to create thin films. This material has a photoluminescence peak at 407 nm with a quantum yield of 26% as nanoplatelets. Single crystals have been grown with a 412 nm emission of and quantum efficiency of 0.22(1). LED devices using this material as the emissive layer have been constructed that opperate at room temperature with an on/off voltage of 2.5-2.8 V and external quantum efficiency (EQE) of 0.038%.

Specifications

CAS 131457-16-4
Molecular Formula C16H24N2PbBr4
Linear Formula C16H24N2PbBr4
Quantity 1 g
Molecular Weight (g/mol) 771.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.