Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(propylene glycol) dimethyl ether, mixture of isomers

Catalog No. 5204621L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-046-21L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-046-21L Supplier Sigma Aldrich Supplier No. 5204621L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aprotic solvent for use on proton-sensitive systems such as water-based polyurethane coatings.Proglyde is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Specifications

CAS 111109-77-4
Density 0.902 g/mL (at 25°C)
Boiling Point 175°C
Molecular Formula C8H18O3
Linear Formula CH3OC3H6OC3H6OCH3
Refractive Index n20/D 1.408 (literature)
Quantity 1 L
Synonym Proglyde™ DMM; Bis(methoxypropyl) ether
Molecular Weight (g/mol) 162.23
Percent Purity ≥99.1%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.