Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(propylene glycol) methyl ether acetate, mixture of isomers

Catalog No. 4065621L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-656-21L 1 L
40-656-2250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-656-21L Supplier Sigma Aldrich Supplier No. 4065621L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Di(propylene glycol) methyl ether acetate (dpma, 3-(3-methoxypropoxy)propyl acetate) is extensively employed as a solvent in coatings and silkscreening inks
  • It is also used as a cleaning agent
  • In vivo, dpma rapidly hydrolyzes to dipropylene glycol methyl ether (dpm), thus, the in vivo effects of dpma are expected to be the same as those of dpm

Specifications

CAS 88917-22-0
Density 0.97 g/mL (at 25°C (literature))
Boiling Point 200°C (lit.)
Molecular Formula C9H18O4
Linear Formula CH3CO2C3H6OC3H6OCH3
Refractive Index n20/D 1.417 (literature)
Quantity 1 L
Molecular Weight (g/mol) 190.24
Percent Purity 99%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.