Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(propylene glycol) methyl ether, mixture of isomers

Catalog No. 2832823L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-328-23L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-328-23L Supplier Sigma Aldrich Supplier No. 2832823L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di(propylene glycol) methyl ether, mixture of isomers

Specifications

CAS 34590-94-8
Density 0.938 g/mL (at 25°C)
Boiling Point 190°C
Molecular Formula C7H16O3
Refractive Index n20/D 1.422 (literature)
Linear Formula CH3OC3H6OC3H6OH
Quantity 3 L
Synonym Dipropylene glycol monomethyl ether
Molecular Weight (g/mol) 148.2
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.